Merck
The Yin and Yang of NMDA receptor signalling.
Trends in neurosciences (2003-01-22)
Giles E Hardingham, Hilmar Bading
ABSTRACT
Ca(2+) entry through the NMDA subtype of glutamate receptors has the power to determine whether neurons survive or die. Too much NMDA receptor activity is harmful to neurons - but so is too little. Is it a case of too much or too little Ca(2+) influx causing cell death or do other factors, such as receptor location or receptor-associated proteins, play a role? Understanding the mechanisms behind this dichotomous signalling is an important area of molecular neuroscience with direct clinical implications.
Sigma-Aldrich
N-Methyl-D-aspartic acid, ≥98% (TLC), solid