Merck
  • Syntheses of N-vanillyl-nonanamide glycosides using amyloglucosidase from Rhizopus and beta-glucosidase from sweet almond.
Syntheses of N-vanillyl-nonanamide glycosides using amyloglucosidase from Rhizopus and beta-glucosidase from sweet almond.
Biotechnology letters (2007-07-05)
Ramaiah Sivakumar, Soundar Divakar
ABSTRACT
Enzymatic syntheses of N-vanillyl-nonanamide, 1, glycosides with D-glucose, 2, D-galactose, 3, D-mannose, 4, D-ribose, 5, maltose, 6, and lactose, 7, were carried out using amyloglucosidase from Rhizopus and beta-glucosidase from sweet almond. The latter catalysed the syntheses of N-vanillyl-nonanamide glycosides (8-13) and exclusively yielded beta-glycosides with carbohydrates 2, 3, 4, 6 and 7, while amyloglucosidase yielded C1-alpha- and beta-glycosides and 6-O-aryl derivatives (8, 9, 11 and 12).
Sigma-Aldrich
Amyloglucosidase from Rhizopus sp., ≥40,000 units/g solid
Pricing and availability is not currently available.