Merck
  • Synthesis and characterization of 1,2-bis(2-(5-bromo-2-hydroxybenzilidenamino)-4-chlorophenoxy)ethane and its metal complexes: an experimental, theoretical, electrochemical, antioxidant and antibacterial study.
Synthesis and characterization of 1,2-bis(2-(5-bromo-2-hydroxybenzilidenamino)-4-chlorophenoxy)ethane and its metal complexes: an experimental, theoretical, electrochemical, antioxidant and antibacterial study.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-10-08)
Salih Ilhan, Haci Baykara, Abdussamet Oztomsuk, Veysi Okumus, Abdulkadir Levent, M Salih Seyitoglu, Sadin Ozdemir
ABSTRACT
A new Schiff base ligand was synthesized by reaction of 5-bromosalicylaldehyde with 1,2-bis(4-chloro-2-aminophenoxy)ethane. Then the Schiff base complexes were synthesized by the reaction of metal salts and the novel Schiff base. The molar conductivity properties of the complexes were studied and found out that the complexes are nonelectrolytes. The structures of the ligand and its metal complexes were characterized by elemental analysis, FT-IR, UV-VIS, magnetic susceptibility measurements, molar conductivity measurements, and thermal gravimetric analysis. In addition antioxidant, theoretical NMR studies and cyclic voltammetry of the complexes were done. Two methods namely metal chelating activity and diphenylpicrylhydrazyl (DPPH) radical scavenging method were used to determine the antioxidant activity, and antibacterial properties of the compounds were also studied.
Sigma-Aldrich
5-Bromosalicylaldehyde, 98%
Pricing and availability is not currently available.