Merck
  • New ultraviolet labelling agents for high-performance liquid chromatographic determination of monocarboxylic acids.
New ultraviolet labelling agents for high-performance liquid chromatographic determination of monocarboxylic acids.
Journal of chromatography (1989-11-03)
K Funazo, M Tanaka, Y Yasaka, H Takigawa, T Shono
ABSTRACT
New UV-labelling agents have been synthesized, which are designed to convert monocarboxylic acids into their highly UV-absorbing derivatives for enhancement of the sensitivities of UV detection in high-performance liquid chromatography. The reagents are p-nitrobenzyl, 3,5-dinitrobenzyl and 2-(phthalimino)ethyl p-toluenesulphonates. Each has been prepared by reaction of p-toluenesulphonyl chloride with p-nitrobenzyl alcohol, 3,5-dinitrobenzyl alcohol or N-(hydroxyethyl)phtalimide, respectively, in the presence of sodium hydroxide, and they are stable in the solid state for at least 6 months. Monocarboxylic acids were derivatized to their p-nitrobenzyl, 3,5-dinitrobenzyl or 2-(phthalimino)ethyl esters with each of the above reagents, respectively, then determined by high-performance liquid chromatography with UV detection. In the UV-labelling with each reagent, 18-crown-6 was used as the catalyst. The effects of the reaction solvent, reaction temperature and time and the concentrations of each reagent and the catalyst were also examined.
Sigma-Aldrich
3,5-Dinitrobenzyl alcohol, 98%
Pricing and availability is not currently available.