Merck
  • Spectroscopic studies on the interaction of sodium benzoate, a food preservative, with calf thymus DNA.
Spectroscopic studies on the interaction of sodium benzoate, a food preservative, with calf thymus DNA.
Food chemistry (2013-06-19)
Guowen Zhang, Yadi Ma
ABSTRACT
The interaction between sodium benzoate (SB) and calf thymus DNA in simulated physiological buffer (pH 7.4) using acridine orange (AO) dye as a fluorescence probe, was investigated by UV-Vis absorption, fluorescence and circular dichroism (CD) spectroscopy along with DNA melting studies and viscosity measurements. An expanded UV-Vis spectral data matrix was resolved by multivariate curve resolution-alternating least squares (MCR-ALS) approach. The equilibrium concentration profiles and the pure spectra for SB, DNA and DNA-SB complex from the high overlapping composite response were simultaneously obtained. The results indicated that SB could bind to DNA, and hydrophobic interactions and hydrogen bonds played a vital role in the binding process. Moreover, SB was able to quench the fluorescence of DNA-AO complex through a static procedure. The quenching observed was indicative of an intercalative mode of interaction between SB and DNA, which was supported by melting studies, viscosity measurements and CD analysis.
Sigma-Aldrich
Deoxyribonucleic acid from calf thymus, Genomic, unsheared
Sigma-Aldrich
Deoxyribonucleic acid from calf thymus, Type XV, Activated, lyophilized powder
Sigma-Aldrich
Acridine Orange hydrochloride solution, 10 mg/mL in H2O, ≥95.0% (HPLC)
Sigma-Aldrich
Acridine Orange solution, 2% in H2O
Sigma-Aldrich
Acridine Orange hydrochloride hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium benzoate, puriss., meets analytical specification of Ph. Eur., BP, FCC, E211, 99.0-100.5% (calc. to the dried substance), powder
Sigma-Aldrich
Deoxyribonucleic acid, single stranded from calf thymus, lyophilized powder
Sigma-Aldrich
Acridine Orange base, Dye content 75 %
Sigma-Aldrich
Sodium benzoate, purum p.a., ≥99.0% (NT)
Sigma-Aldrich
Sodium benzoate, BioXtra, ≥99.5%
Supelco
Sodium benzoate, Pharmaceutical Secondary Standard; Certified Reference Material