Merck
  • Carboplatin plus either docetaxel or paclitaxel for Japanese patients with advanced non-small cell lung cancer.
Carboplatin plus either docetaxel or paclitaxel for Japanese patients with advanced non-small cell lung cancer.
Anticancer research (2013-10-15)
Masaaki Kawahara, Shinji Atagi, Kiyoshi Komuta, Hiroshige Yoshioka, Masayuki Kawasaki, Yuka Fujita, Toshiro Yonei, Fumitaka Ogushi, Kaoru Kubota, Naoyuki Nogami, Michiko Tsuchiya, Kazuhiko Shibata, Yoshio Tomizawa, Koichi Minato, Kazuya Fukuoka, Kazuhiro Asami, Takeharu Yamanaka
ABSTRACT
Assessment of the efficacy of docetaxel plus carboplatin vs. paclitaxel plus carboplatin in Japanese patients with advanced non-small cell lung cancer (NSCLC). Chemotherapy-naïve patients were randomly assigned at a ratio of 2 to 1 to receive six cycles of either docetaxel (60 mg/m(2)) plus carboplatin [area under the curve (AUC)=6 mg/ml min] or paclitaxel (200 mg/m(2)) plus carboplatin (same dose), on day 1 every 21 days. The primary end-point was progression-free survival (PFS). A total of 90 patients were enrolled. Overall response rate, median PFS and median survival time in the docetaxel-plus-carboplatin group and the paclitaxel-plus-carboplatin group were 23% vs. 33%, 4.8 months vs. 5.1 months, and 17.6 months vs. 15.6 months, respectively. The docetaxel-plus-carboplatin group had a higher incidence of grade 3 or 4 neutropenia (88% vs. 60%). Both regimens were similarly effective in Japanese patients with advanced NSCLC.
Sigma-Aldrich
Carboplatin