Merck
  • Analysis of omeprazole and 5-OH omeprazole in human plasma using hydrophilic interaction chromatography with tandem mass spectrometry (HILIC-MS/MS)--eliminating evaporation and reconstitution steps in 96-well liquid/liquid extraction.
Analysis of omeprazole and 5-OH omeprazole in human plasma using hydrophilic interaction chromatography with tandem mass spectrometry (HILIC-MS/MS)--eliminating evaporation and reconstitution steps in 96-well liquid/liquid extraction.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2005-11-22)
Qi Song, Weng Naidong
ABSTRACT
Bioanalytical methods using liquid/liquid extraction (LLE) and liquid chromatography with electrospray tandem mass spectrometry (LC-MS/MS) are widely used. The organic extracts need to be evaporated and reconstituted, hampering further improvement of throughput and automation. In this study, we demonstrated a novel approach of eliminating these two steps in 96-well LLE by using hydrophilic interaction chromatography with MS/MS (HILIC-MS/MS) on silica column with high organic/low aqueous mobile phase. Omeprazole, its metabolite 5-OH omeprazole, and internal standard desoxyomeprazole, were extracted from 0.05 ml of human plasma using 0.5 ml of ethyl acetate in a 96-well plate. A portion (0.1 ml) of the ethyl acetate extract was diluted with 0.4 ml of acetonitrile and 10 microl was injected onto a Betasil silica column (50 mm x 3.0 mm, 5 microm) and detected by API 3000 and 4000 with (+) ESI. Mobile phase with linear gradient elution consists of acetonitrile, water, and formic acid (from 95:5:0.1 to 73.5:26.5:0.1 in 2 min). The flow rate was 1.5 ml/min with total run time of 2.75 min. The method was validated for a low limit of quantitation at 2.5 ng/ml for both analytes. The method was also validated for specificity, reproducibility, stability and recovery. Lack of adverse matrix effect and carry-over was also demonstrated. The inter-day precision and accuracy of the quality control samples at low, medium and high concentration levels were <4.4% relative standard deviation (R.S.D.) and 4.1% relative error (R.E.) for omeprazole, and 4.5% R.S.D. and 5.6% R.E. for 5-OH omeprazole, respectively.
Product Description
Supelco
5-Hydroxyomeprazole, analytical standard