Merck
  • HPLC analysis of the second-generation antidepressant sertraline and its main metabolite N-desmethylsertraline in human plasma.
HPLC analysis of the second-generation antidepressant sertraline and its main metabolite N-desmethylsertraline in human plasma.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2006-04-04)
Roberto Mandrioli, Maria Addolorata Saracino, Silvia Ferrari, Domenico Berardi, Ernst Kenndler, Maria Augusta Raggi
ABSTRACT
A liquid chromatographic method with ultraviolet detection was developed for the analysis of the recent antidepressant sertraline and its main metabolite N-desmethylsertraline in human plasma. The analytes were separated on a C8 reversed phase column, using a mobile phase composed of acetonitrile and a 12.3 mM, pH 3.0 phosphate buffer containing 0.1% triethylamine (35:65, v/v). Clomipramine was used as the Internal Standard. Using a solid phase extraction procedure with C2 cartridges high extraction yields (>94%) and good purification from matrix interference were obtained. Good linearity was obtained in the 7.5-250.0 ng mL(-1) range for sertraline and in the 10-500 ng mL(-1) range for N-desmethylsertraline. The analytical method was validated in terms of precision, extraction yield and accuracy. These assays gave R.S.D.% values for precision always lower than 3.9% and mean accuracy higher than 90%. Thanks to its good selectivity, the method proved to be suitable for the analysis of plasma samples from patients treated with sertraline as either monotherapy or polypharmacy.
Product Description
Supelco
(±)-Norsertraline hydrochloride solution, 100 μg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Pricing and availability is not currently available.