- Hydrogen physisorption in high SSA microporous materials - A comparison between AX-2133 and MOF-177 at cryogenic conditions
Hydrogen physisorption in high SSA microporous materials - A comparison between AX-2133 and MOF-177 at cryogenic conditions
International Journal of Hydrogen Energy (2011)
Schlichtenmayer, et al.