Merck
  • Trapping efficiency of polyurethane foam and Amberlite XAD-2 for various organochlorine compounds
Trapping efficiency of polyurethane foam and Amberlite XAD-2 for various organochlorine compounds
Fresenius Journal of Analytical Chemistry (1995)
Nerin, C., et al.