Merck
  • Mono-or narrow disperse poly (methacrylate-co-divinylbenzene) microspheres by precipitation polymerization
Mono-or narrow disperse poly (methacrylate-co-divinylbenzene) microspheres by precipitation polymerization
Journal of Polymer Science Part A: Polymer Chemistry (1999)
Li, W. H., & Stover, H. D.