- Development of branching in atom transfer radical copolymerization of styrene with triethylene glycol dimethacrylate
Development of branching in atom transfer radical copolymerization of styrene with triethylene glycol dimethacrylate
Macromolecules (2009)
Yang, H. J., Jiang, B. B., Huang, W. Y., Zhang, D. L., Kong, L. Z., Chen, J. H., & Yang, Y.