Merck
  • Supplementation of Bolton broth with triclosan improves detection of Campylobacter jejuni and Campylobacter coli in chicken carcass rinse.
Supplementation of Bolton broth with triclosan improves detection of Campylobacter jejuni and Campylobacter coli in chicken carcass rinse.
International journal of food microbiology (2014-05-13)
Jung-Whan Chon, Young-Ji Kim, Hong-Seok Kim, Dong-Hyeon Kim, Hyunsook Kim, Kwang-Young Song, Kun-Ho Seo
ABSTRACT
We compared Bolton enrichment broth supplemented with antimicrobial triclosan (T-Bolton broth) and normal Bolton broth for the isolation of Campylobacter jejuni (C. jejuni) and Campylobacter coli (C. coli) from chicken carcass rinse. Whole chickens were rinsed with buffered peptone water prior to enrichment in normal Bolton broth or T-Bolton broth, followed by inoculation onto modified charcoal-cefoperazone-deoxycholate agar (mCCDA). Suspect colonies were confirmed by PCR. We observed a significantly higher number of C. jejuni or C. coli-positive samples in the T-Bolton broth (71.3%) than in the normal Bolton broth (27.5%) (p<0.05). Furthermore, the number of contaminated mCCDA plates was lower after enrichment in T-Bolton broth (3.8%) than in the normal Bolton broth (75%) (p<0.05), indicating that T-Bolton broth has higher selectivity. Finally, we identified extended-spectrum β-lactamase-producing Escherichia coli as the predominant competing flora in normal Bolton broth. In conclusion, the use of T-Bolton broth results in significant elimination of competing bacteria.
Product Description
Supelco
Triclosan, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Triclosan, 97.0-103.0% (active substance, GC)