Merck
  • Charcot-Marie-Tooth neuropathy type 1B is associated with mutations of the myelin P0 gene.
Charcot-Marie-Tooth neuropathy type 1B is associated with mutations of the myelin P0 gene.
Nature genetics (1993-09-01)
K Hayasaka, M Himoro, W Sato, G Takada, K Uyemura, N Shimizu, T D Bird, P M Conneally, P F Chance
ABSTRACT
P0, a major structural protein of peripheral myelin, is a homophilic adhesion molecule and maps to chromosome 1q22-q23, in the region of the locus for Charcot-Marie-Tooth neuropathy type 1B (CMT1B). We have investigated P0 as a candidate gene in two pedigrees with CMT1B and found point mutations which are completely linked with the disease (Z = 5.5, theta = 0). The mutations, glutamate substitution for lysine 96 or aspartate 90, are located in the extracellular domain, which plays a significant role in myelin membrane adhesion. Individuals with CMT1B are heterozygous for the normal allele and the mutant allele. Our results indicate that P0 is a gene responsible for CMT1B.