Merck
  • Measuring localization and diffusion coefficients of basolateral proteins in lateral versus basal membranes using functionalized substrates and kICS analysis.
Measuring localization and diffusion coefficients of basolateral proteins in lateral versus basal membranes using functionalized substrates and kICS analysis.
Biochimica et biophysica acta (2014-06-21)
Saw Marlar, Eva C Arnspang, Gitte A Pedersen, Jennifer S Koffman, Lene N Nejsum
ABSTRACT
Micropatterning enabled semiquantitation of basolateral proteins in lateral and basal membranes of the same cell. Lateral diffusion coefficients of basolateral aquaporin-3 (AQP3-EGFP) and EGFP-AQP4 were extracted from "lateral" and "basal" membranes using identical live-cell imaging and k-space Image Correlation Spectroscopy (kICS). To simultaneously image proteins in "lateral" and "basal" membranes, micropatterning with the extracellular domain of E-cadherin and collagen, to mimic cell-cell and cell-extracellular matrix (ECM) adhesion, respectively, was used. In kidney collecting duct principal cells AQP3 localizes lateral and basal whereas AQP4 localizes mainly basal. On alternating stripes of E-cadherin and collagen, AQP3-EGFP was predominantly localized to "lateral" compared to "basal" membranes, whereas Orange-AQP4 was evenly distributed. Average diffusion coefficients were extracted via kICS analysis of rapid time-lapse sequences of AQP3-EGFP and EGFP-AQP4 on uniform substrates of either E-cadherin or collagen. AQP3-EGFP was measured to 0.022±0.010μm(2)/s on E-cadherin and 0.019±0.004μm(2)/s on collagen, whereas EGFP-AQP4 was measured to 0.044±0.009μm(2)/s on E-cadherin and 0.037±0.009μm(2)/s on collagen, thus, diffusion did not differ between substrates. Cholesterol depletion by methyl-beta-cyclodextrin (MBCD) reduced the AQP3-EGFP diffusion coefficient by 43% from 0.024±0.007μm(2)/s (water) to 0.014±0.003μm(2)/s (MBCD) (p<0.05) on collagen surfaces, and by 41% from 0.023±0.011μm(2)/s (water) to 0.014±0.005μm(2)/s (MBCD) (p<0.05) on E-cadherin surfaces. Thus, protein patterning enables the semiquantitation of protein distribution between the "lateral" and "basal" membranes as well as measurements of lateral diffusion coefficients.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material