Merck
  • Anionic polymerization of carbazolyl-substituted oxiranes initiated by potassium alkalide, potassium tert-butoxide and potassium hydride.
Anionic polymerization of carbazolyl-substituted oxiranes initiated by potassium alkalide, potassium tert-butoxide and potassium hydride.
Macromolecular Chemistry and Physics (1995)
Buika G, et al.