Merck
  • Rapid, specific, and sensitive measurements of plasma sphingomyelin and phosphatidylcholine.
Rapid, specific, and sensitive measurements of plasma sphingomyelin and phosphatidylcholine.
Journal of lipid research (2005-12-24)
Mohammad Reza Hojjati, Xian-Cheng Jiang
ABSTRACT
Sphingomyelin (SM) and phosphatidylcholine (PC) are two major phospholipids on plasma lipoproteins. Their concentration is classically measured by lipid extraction, thin-layer chromatography, and phosphate determination on separated SM or PC spots. Here, we describe two rapid, specific, and sensitive enzymatic measurements for both phospholipids. Plasma was incubated with bacterial sphingomyelinase (for SM measurement) or bacterial PC-specific phospholipase D (for PC measurement), alkaline phosphatase, choline oxidase, peroxidase, N-ethyl-N-(2-hydroxy-3-sulfopropyl)-3,5-dimethoxyaniline, and 4-aminoantipyrine for 45 min. A blue dye, with an optimal absorption at 595 nm, was generated. PC levels did not influence SM measurement and vice versa. The linear range for the SM measurement was 0.5-5 microg, and that for PC was 2.5-20 microg. The mean percentage recovery was 98.0 +/- 5.2% for SM and 96.6 +/- 3.8% for PC. The interassay coefficient of variation of the assay was 1.7 +/- 0.05% for SM and 3.1 +/- 0.13% for PC. These two new methods are amenable to automation and can be adapted for large-scale, high-throughput assays.
Sigma-Aldrich
Sphingomyelin, from chicken egg yolk, ≥95%
Pricing and availability is not currently available.