Merck
  • Vitamin K nutritional status and undercarboxylated osteocalcin in postmenopausal osteoporotic women treated with bisphosphonates.
Vitamin K nutritional status and undercarboxylated osteocalcin in postmenopausal osteoporotic women treated with bisphosphonates.
Asia Pacific journal of clinical nutrition (2014-06-06)
Jun Iwamoto, Tetsuya Takada, Yoshihiro Sato
ABSTRACT
Serum undercarboxylated osteocalcin (ucOC) is an index of vitamin K nutritional status in treatment-naive postmenopausal osteoporotic women. The purpose of the present study was to reveal the association between vitamin K nutritional status and serum ucOC concentrations in postmenopausal osteoporotic women taking bisphosphonates. Eighty-six postmenopausal women with osteoporosis (age range: 47-90 years) initiated bisphosphonate treatment. Vitamin K nutritional status was evaluated using a simple vitamin K-intake questionnaire and serum ucOC concentrations were measured after 6 months of treatment. The patients were divided into two groups according to the simple vitamin K-intake questionnaire score: a low vitamin K-intake (score <40) group (n=67) and a normal vitamin K-intake (score >=40) group (n=19). There were no significant differences between the groups in baseline parameters including age, height, body weight, body mass index, serum alkaline phosphatase (ALP), urinary cross-linked N-terminal telopeptides of type I collagen (NTX), and changes in serum ALP and urinary NTX concentrations during the 6-month treatment period. However, the mean serum ucOC concentration after 6 months of treatment was significantly higher in the low vitamin K-intake group (2.79 ng/mL) than in the normal vitamin K-intake group (2.20 ng/mL). These results suggest that 78% of postmenopausal osteoporotic women treated with bisphosphonates may have vitamin K deficiency as indicated by low vitamin K-intake and high serum ucOC concentrations, despite having a similar reduction in bone turnover to women who have normal vitamin K-intake.
Sigma-Aldrich
Ethyl octanoate, ≥98%, FCC, FG
Pricing and availability is not currently available.