Merck
PLAGL2 regulates actin cytoskeletal architecture and cell migration.
Carcinogenesis (2014-03-29)
Ryuichiro Sekiya, Masao Maeda, Hong Yuan, Eri Asano, Toshinori Hyodo, Hitoki Hasegawa, Satoko Ito, Kiyosumi Shibata, Michinari Hamaguchi, Fumitaka Kikkawa, Hiroaki Kajiyama, Takeshi Senga
ABSTRACT
Pleomorphic adenoma gene like-2 (PLAGL2), a member of the PLAG gene family, is a C2H2 zinc finger transcriptional factor that is involved in cellular transformation and apoptosis. In this report, we show that PLAGL2 is associated with the organization of stress fibers and with small guanosine triphosphatase (GTPase) activity. Depletion of PLAGL2 in two different ovarian cancer cell lines, ES-2 and HEY, induced activation of RhoA, whereas activity of Rac1 was suppressed. Organization of actin stress fibers and focal adhesions was significantly promoted by PLAGL2 knockdown in a RhoA-dependent manner. Conversely, exogenous expression of PLAGL2 in MDA-MB-231 cells, a breast cancer cell line, resulted in the activation of Rac1 and the inactivation of RhoA. In addition, PLAGL2 expression induced lamellipodia formation and disruption of stress fiber formation. Finally, we show that CHN1 expression is essential for Rac1 inactivation in PLAGL2-depleted cells. Our results demonstrate a crucial role of PLAGL2 in actin dynamics and give further insight into the role of PLAGL2 in cellular transformation and apoptosis.
Product Description
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous