- Molecular Structures of Hydrogen Cyanide and Acetonitrile as Studied by Gas Electron Diffraction
Molecular Structures of Hydrogen Cyanide and Acetonitrile as Studied by Gas Electron Diffraction
Bulletin of the Chemical Society of Japan (1974)
Karakida K, et al.