Merck
  • Electrogenerated chemiluminescent emission from an organized (L-B) monolayer of a Ru(bpy)32+-based surfactant on semiconductor and metal electrodes.
Electrogenerated chemiluminescent emission from an organized (L-B) monolayer of a Ru(bpy)<SUB>3</SUB><SUP>2+</SUP>-based surfactant on semiconductor and metal electrodes.
The Journal of Physical Chemistry (1988)
Zhang X and Bard AJ.