Merck
  • Evaluation of Adhesion between Nafion and gold electrodes fabricated through novel dry process utilizing argon plasma treatment.
Evaluation of Adhesion between Nafion and gold electrodes fabricated through novel dry process utilizing argon plasma treatment.
Chemistry Letters (Jpn) (2013)
Hiroshiba N, et al.