Merck
  • YAP1 is a potential biomarker for cetuximab resistance in head and neck cancer.
YAP1 is a potential biomarker for cetuximab resistance in head and neck cancer.
Oral oncology (2014-07-06)
Fredrik Jerhammar, Ann-Charlotte Johansson, Rebecca Ceder, Jenny Welander, Agneta Jansson, Roland C Grafström, Peter Söderkvist, Karin Roberg
ABSTRACT
Targeted therapy against the epidermal growth factor receptor (EGFR) only variably represents a therapeutic advance in head and neck squamous cell carcinoma (HNSCC). This study addresses the need of biomarkers of treatment response to the EGFR-targeting antibody cetuximab (Erbitux®). The intrinsic cetuximab sensitivity of HNSCC cell lines was assessed by a crystal violet assay. Gene copy number analysis of five resistant and five sensitive cell lines was performed using the Affymetrix SNP 6.0 platform. Quantitative real-time PCR was used for verification of selected copy number alterations and assessment of mRNA expression. The functional importance of the findings on the gene and mRNA level was investigated employing siRNA technology. The data was statistically evaluated using Mann-Whitney U-test and Spearman's correlation test. Analysis of the intrinsic cetuximab sensitivity of 32 HNSCC cell lines characterized five and nine lines as cetuximab sensitive or resistant, respectively. Gene copy number analysis of five resistant versus five sensitive cell lines identified 39 amplified protein-coding genes, including YAP1, in the genomic regions 11q22.1 or 5p13-15. Assessment using qPCR verified that YAP1 amplification associated with cetuximab resistance. Amplification of YAP1 correlated to higher mRNA levels, and RNA knockdown resulted in increased cetuximab sensitivity. Assessment of several independent clinical data sets in the public domain confirmed YAP1 amplifications in multiple tumor types including HNSCC, along with highly differential expression in a subset of HNSCC patients. Taken together, we provide evidence that YAP1 could represent a novel biomarker gene of cetuximab resistance in HNSCC cell lines.
Sigma-Aldrich
Human EGFR / ErbB1 ELISA Kit, for serum, plasma, cell culture supernatants and urine
Pricing and availability is not currently available.
Sigma-Aldrich
Phosphotyrosine EGFR / Epidermal Growth Factor Receptor ELISA Kit
Pricing and availability is not currently available.