Merck
  • Synthesis, biological activity, and biopharmaceutical characterization of tacrine dimers as acetylcholinesterase inhibitors.
Synthesis, biological activity, and biopharmaceutical characterization of tacrine dimers as acetylcholinesterase inhibitors.
International journal of pharmaceutics (2014-12-03)
Shuai Qian, Lisi He, Marvin Mak, Yifan Han, Chun-Yu Ho, Zhong Zuo
ABSTRACT
Tacrine (THA), as the first approved acetylcholinesterase (AChE) inhibitors for the treatment of Alzheimer's disease (AD), has been extensively investigated in last seven decades. After dimerization of THA via a 7-carbon alkyl spacer, bis(7)-tacrine (B7T) showed much potent anti-AChE activity than THA. We here report synthesis, biological evaluation and biopharmaceutical characterization of six THA dimers referable to B7T. According to IC50 values, the in vitro anti-AChE activities of THA dimers were up to 300-fold more potent and 200-fold more selective than that of THA. In addition, the anti-AChE activities of THA dimers were found to be associated with the type and length of the linkage. All studied THA dimers showed much lower cytotoxicity than B7T, but like B7T, they demonstrated much lower absorptive permeabilities than that of THA on Caco-2 monolayer model. In addition, all THA dimers demonstrated significant efflux transport (efflux ratio >4), indicating that the limited permeability could be associated with the efflux transport during absorption process. Moreover, the dimer with higher Log P value was accompanied with higher permeability but lower aqueous solubility. A balanced consideration of activity, solubility, cytotoxicity and permeability should be conducted in selection of the potential candidates for further in vivo investigation.
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
9-Amino-1,2,3,4-tetrahydroacridine hydrochloride hydrate, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
9-Amino-1,2,3,4-tetrahydroacridine hydrochloride hydrate, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Sodium phosphate dibasic solution, BioUltra, 0.5 M in H2O
Supelco
Phosphate Standard for IC, TraceCERT®, 1000 mg/L phosphate in water (nominal concentration)
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Supelco
Triethylamine, analytical standard
Sigma-Aldrich
Sodium phosphate dibasic, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Sodium phosphate dibasic, purum p.a., anhydrous, ≥98.0% (T)
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, Molecular Biology, anhydrous, ≥98%
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, Molecular Biology, ≥98.5% (titration)
Sigma-Aldrich
Sodium phosphate dibasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium phosphate dibasic, 99.95% trace metals basis
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Sodium phosphate dibasic, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%, free-flowing, Redi-Dri
Sigma-Aldrich
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%