Merck
  • Comparative analysis of hapalindole, ambiguine and welwitindolinone gene clusters and reconstitution of indole-isonitrile biosynthesis from cyanobacteria.
Comparative analysis of hapalindole, ambiguine and welwitindolinone gene clusters and reconstitution of indole-isonitrile biosynthesis from cyanobacteria.
BMC microbiology (2014-09-10)
Melinda L Micallef, Deepti Sharma, Brittney M Bunn, Lena Gerwick, Rajesh Viswanathan, Michelle C Moffitt
ABSTRACT
The hapalindole-type family of natural products is a group of hybrid isoprenoid-indole alkaloids, produced solely by members of the Subsection V cyanobacterial strains. This family broadly includes the hapalindoles, welwitindolinones, fisherindoles and ambiguines amongst others, all of which have an isonitrile- or isothiocyanate-containing indole alkaloid skeleton, with a cyclized isoprene unit. The hapalindoles are diversified into the welwitindolinones, fischerindoles and ambiguines through the employment of tailoring oxygenase, methyltransferase and prenyltransferase enzymes. We compare the genetic basis for the biosynthesis of this diverse group of natural products and identify key early biosynthetic intermediates. Whole genome sequencing of freshwater and terrestrial cyanobacteria Westiella intricata UH strain HT-29-1, Hapalosiphon welwitschii UH strain IC-52-3, Fischerella ambigua UTEX 1903 and Fischerella sp. ATCC 43239 led to the identification of a candidate hapalindole-type gene cluster in each strain. These were compared with the recently published ambiguine and welwitindolinone gene clusters and four unpublished clusters identified within publicly available genomes. We present detailed comparative bioinformatic analysis of the gene clusters and the biosynthesis of a pivotal indole-isonitrile intermediate resulting in both cis and trans geometrical isomers. Enzyme analyses and metabolite extractions from two hapalindole-producing Fischerella strains indicate the presence of cis and trans indole-isonitriles as biosynthetic intermediates in the early steps of the pathway. Interestingly, the organization of the welwitindolinone gene cluster is conserved in all producing strains but distinct from the hapalindole and ambiguine clusters. Enzymatic assays using WelI1 and WelI3 from Westiella intricata UH strain HT-29-1 demonstrated the ability to catalyze the formation of both cis and trans geometrical isomers when using a cell lysate. The enzymatic and metabolic characterization of both cis and trans indole-isonitrile intermediates implies conservation of their stereochemical integrity towards members of the ambiguine and welwitindolinone products. In summary, we present data that supports a unified biosynthetic pathway towards hapalindoles in nine individual species of cyanobacteria. Diversification of the pathway occurs later through the employment of specialized enzymatic steps towards fischerindoles, ambiguines and welwitindolinones.
Product Description
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Imidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Ondansetron impurity E, European Pharmacopoeia (EP) Reference Standard
USP
Imidazole, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Imidazole, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Imidazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Imidazole, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Imidazole, Molecular Biology, ≥99% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%, Redi-Dri, free-flowing
Supelco
Glycerol, analytical standard
Sigma-Aldrich
Glycerol, Vetec, reagent grade, 99%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Sigma-Aldrich
Imidazole buffer Solution, BioUltra, 1 M in H2O
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)