Merck
  • Helicobacter pylori modulates cisplatin sensitivity in gastric cancer by down-regulating miR-141 expression.
Helicobacter pylori modulates cisplatin sensitivity in gastric cancer by down-regulating miR-141 expression.
Helicobacter (2014-03-19)
Xiaoying Zhou, Jing Su, Li Zhu, Guoxin Zhang
ABSTRACT
Recent studies found that gastric cancer patients with Helicobacter pylori infection had a better response to chemotherapy and had an improved overall prognosis compared with those without. However, the underlying mechanism remains unknown. Quantitative real-time PCR (qRT-PCR) was utilized to determine the expression profile of miR-141 in H. pylori infected cells and tissues and their respective controls. qRT-PCR and Western blot were used to determine the expression level of KEAP-1. Luciferase reporter assays were used to determine whether KEAP-1 was a direct target of miR-141 in the gastric cancer cells. MTT and apoptosis assay were performed to detect the survival of cells under cisplatin treatment. We found that H. pylori infection can significantly down-regulate miR-141 expression. Knockdown miR-141 expression in 7901/DDP and 7901 cells could significantly improve cisplatin sensitivity. Over-expression of miR-141 resulted in enhanced resistance to cisplatin in both gastric cancer cells. We also demonstrated that miR-141 directly targets KEAP1 by luciferase reporter assay, and that down-regulation of KEAP1 induces cisplatin resistance. Conversely, over-expression of KEAP1 significantly enhanced cisplatin sensitivity. Our 75 pairs of tissues also showed that KEAP1 was significantly up-regulated in H. pylori-positive tissues. Altogether, these findings demonstrated that the H. pylori infection could modulate cisplatin sensitivity through miR-141-mediated regulation of KEAP1.
Sigma-Aldrich
Anti-STAT4 (Ab-693) antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-STAT4 antibody produced in rabbit, IgG fraction of antiserum
Pricing and availability is not currently available.