Merck
  • In situ normothermic regional perfusion for controlled donation after circulatory death--the United Kingdom experience.
In situ normothermic regional perfusion for controlled donation after circulatory death--the United Kingdom experience.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons (2014-10-07)
G C Oniscu, L V Randle, P Muiesan, A J Butler, I S Currie, M T P R Perera, J L Forsythe, C J E Watson
ABSTRACT
Organs recovered from donors after circulatory death (DCD) suffer warm ischemia before cold storage which may prejudice graft survival and result in a greater risk of complications after transplant. A period of normothermic regional perfusion (NRP) in the donor may reverse these effects and improve organ function. Twenty-one NRP retrievals from Maastricht category III DCD donors were performed at three UK centers. NRP was established postasystole via aortic and caval cannulation and maintained for 2 h. Blood gases and biochemistry were monitored to assess organ function. Sixty-three organs were recovered. Forty-nine patients were transplanted. The median time from asystole to NRP was 16 min (range 10-23 min). Thirty-two patients received a kidney transplant. The median cold ischemia time was 12 h 30 min (range 5 h 25 min-18 h 22 min). The median creatinine at 3 and 12 months was 107 µmol/L (range 72-222) and 121 µmol/L (range 63-157), respectively. Thirteen (40%) recipients had delayed graft function and four lost the grafts. Eleven patients received a liver transplant. The first week median peak ALT was 389 IU/L (range 58-3043). One patient had primary nonfunction. Two combined pancreas-kidney transplants, one islet transplant and three double lung transplants were performed with primary function. NRP in DCD donation facilitates organ recovery and may improve short-term outcomes.
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder