Merck
  • Volatile compounds and antioxidant capacity of the bio-oil obtained by pyrolysis of Japanese red pine (pinus densiflora siebold and zucc.).
Volatile compounds and antioxidant capacity of the bio-oil obtained by pyrolysis of Japanese red pine (pinus densiflora siebold and zucc.).
Molecules (Basel, Switzerland) (2015-03-05)
Jayanta Kumar Patra, Sung Hong Kim, Hyewon Hwang, Joon Weon Choi, Kwang-Hyun Baek
ABSTRACT
In the present study, sawdust bio-oil (SBO) manufactured by fast pyrolysis of Japanese red pine (Pinus densiflora Siebold and Zucc.) sawdust was analyzed for its volatile chemical compound composition and evaluated for its free radical scavenging potential, inhibition of lipid peroxidation and reducing power. Gas chromatography and mass spectroscopy revealed 29 volatile compounds, comprising 97.6% of the total volatile compounds in SBO. The antioxidant potential of SBO in terms of IC50 values was 48.44 µg/mL for hydroxyl radical scavenging, 89.52 µg/mL for 1,1-diphenyl-2-picrylhydraxyl radical scavenging, 94.23 µg/mL for 2,2'-azino-bis[3-ethylbenzothiazoline-6-sulphonic acid] radical scavenging, and 136.06 µg/mL for superoxide radical scavenging activity. The total phenol content in SBO was 5.7% gallic acid equivalent. Based on the composition of its volatile compounds, high free radical scavenging potential and antioxidant properties, SBO could be used as a source of antioxidant compounds, flavoring agents and nutraceuticals in the food, pharmaceutical, and cosmetic industries.
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Iron(III) chloride, anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Iron(III) chloride, ≥99.9% trace metals basis
Sigma-Aldrich
Iron(III) chloride solution, 0.2 M in 2-methyltetrahydrofuran
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Millipore
TDA Reagent, suitable for microbiology
Sigma-Aldrich
Phenazine, 98%
Sigma-Aldrich
1,1-Diphenyl-2-picrylhydrazine, 97%
Sigma-Aldrich
Phenazine methosulfate
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Gallic acid, 97.5-102.5% (titration)
Supelco
Gallic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Iron(III) chloride solution, purum, 45% FeCl3 basis
Supelco
Sodium Nitroprusside, Pharmaceutical Secondary Standard; Certified Reference Material