Merck
  • BCOR-CCNB3 fusions are frequent in undifferentiated sarcomas of male children.
BCOR-CCNB3 fusions are frequent in undifferentiated sarcomas of male children.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc (2014-11-02)
Tricia L Peters, Vijetha Kumar, Sumanth Polikepahad, Frank Y Lin, Stephen F Sarabia, Yu Liang, Wei-Lien Wang, Alexander J Lazar, HarshaVardhan Doddapaneni, Hsu Chao, Donna M Muzny, David A Wheeler, M Fatih Okcu, Sharon E Plon, M John Hicks, Dolores López-Terrada, D Williams Parsons, Angshumoy Roy
ABSTRACT
The BCOR-CCNB3 fusion gene, resulting from a chromosome X paracentric inversion, was recently described in translocation-negative 'Ewing-like' sarcomas arising in bone and soft tissue. Genetic subclassification of undifferentiated unclassified sarcomas may potentially offer markers for reproducible diagnosis and substrates for therapy. Using whole transcriptome paired-end RNA sequencing (RNA-seq) we unexpectedly identified BCOR-CCNB3 fusion transcripts in an undifferentiated spindle-cell sarcoma. RNA-seq results were confirmed through direct RT-PCR of tumor RNA and cloning of the genomic breakpoints from tumor DNA. Five additional undifferentiated sarcomas with BCOR-CCNB3 fusions were identified in a series of 42 pediatric and adult unclassified sarcomas. Genomic breakpoint analysis demonstrated unique breakpoint locations in each case at the DNA level even though the resulting fusion mRNA was identical in all cases. All patients with BCOR-CCNB3 sarcoma were males diagnosed in mid childhood (7-13 years of age). Tumors were equally distributed between axial and extra-axial locations. Five of the six tumors were soft-tissue lesions with either predominant spindle-cell morphology or spindle-cell areas interspersed with ovoid to round cells. CCNB3 immunohistochemistry showed strong nuclear positivity in five tumors before oncologic therapy, but was patchy to negative in post-treatment tumor samples. An RT-PCR assay developed to detect the fusion transcript in archival formalin-fixed tissue was positive in all six cases, with high sensitivity and specificity in both pre- and post-treated samples. This study adds to recent reports on the clinicopathologic spectrum of BCOR-CCNB3 fusion-positive sarcomas, a newly emerging entity within the undifferentiated unclassified sarcoma category and describes a simple RT-PCR assay that in conjunction with CCNB3 immunohistochemistry can be useful in diagnosing these tumors.
Sigma-Aldrich
Anti-CCNB3 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.