Merck
  • Preparation of solid dispersion of dronedarone hydrochloride with Soluplus(®) by hot melt extrusion technique for enhanced drug release.
Preparation of solid dispersion of dronedarone hydrochloride with Soluplus(®) by hot melt extrusion technique for enhanced drug release.
Chemical & pharmaceutical bulletin (2015-04-04)
Sang Duk Han, Sang Won Jung, Sun Woo Jang, Hyuck Jun Jung, Miwon Son, Byoung Moon Kim, Myung Joo Kang
ABSTRACT
In order to enhance the dissolution rate of dronedarone hydrochloride (DRN), a novel Soluplus(®) (polyethyleneglycol-polyvinyl caprolactam-polyvinyl acetate grafted copolymer)-based solid dispersion (SD) was formulated using a hot melt extrusion technique. The physical characteristics determined using scanning electron microscopy and X-ray powder diffraction, revealed that the active compound was molecularly dispersed in the amphiphilic polymer in a stable amorphous form. The dissolution rate of DRN from the tablet dosage form of SD extrudate consisted of the drug and Soluplus(®) in a weight ratio of 1 : 1, and was obviously more rapid and higher than that of the intact drug and marketed product (Multaq(®), Sanofi, U.S.A.) at pH 1.2, 4.0 and 6.8. This suggests that Soluplus(®)-based SD formula can be a promising approach for enhancing the dissolution and oral absorption of DRN with a simple preparation process.
Sigma-Aldrich
Sodium hydroxyacetate, AldrichCPR