Merck
  • Development and evaluation of IgY ImmunoCapture PCR ELISA for detection of Staphylococcus aureus enterotoxin A devoid of protein A interference.
Development and evaluation of IgY ImmunoCapture PCR ELISA for detection of Staphylococcus aureus enterotoxin A devoid of protein A interference.
Journal of immunological methods (2014-06-20)
Prakash Reddy, Shylaja Ramlal, Murali Harishchandra Sripathy, Harsh Vardhan Batra
ABSTRACT
In the present study, a sensitive and specific IgY mediated ImmunoCapture-PCR-ELISA (IC-PCR-ELISA) was developed for the detection of staphylococcal enterotoxin A (SEA) from culture supernatants and suspected contaminated samples. Due to the virtue of avian immunoglobulins (IgY) to have the least affinity towards staphylococcal protein A (SpA) responsible for false positives, we employed anti-SEA IgY for capture of SEA toxin and revealed with SEA specific rabbit antibodies conjugated to a 524bp DNA marker. Biotin-11-dUTP was incorporated during PCR amplification and post PCR analysis was performed by PCR-ELISA. Unlike IgG immunocapture, IgY mediated immunocapture of SEA was free from false positives due to protein A. The developed assay was specific to SEA except for minor cross reactivity with staphylococcal enterotoxin E (SEE). Several raw milk samples were evaluated for the presence of SEA with and without enrichment. Three samples were found to be positive for SEA after enrichment for 8h. Though IC-PCR-ELISA for SEA showed 100% correlation with PCR analysis for sea gene, the assay was unique in terms of sensitivity of detecting ~10pg/ml of SEA toxin from spiked milk samples. Result of IC-PCR-ELISA was further confirmed by conventional methods of isolation and characterization. The presented method can be very useful for rapid analysis of milk samples for SEA contamination and can be further extended for detection of multiple SE's in different wells of same PCR plate using common DNA substrate.
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Isopropyl β-D-1-thiogalactopyranoside, ≥99% (TLC)
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Urea, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Isopropyl β-D-thiogalactopyranoside solution, ReadyMade IPTG solution for Blue-white screening
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
USP
Urea, United States Pharmacopeia (USP) Reference Standard
Supelco
Urea, analytical standard
Sigma-Aldrich
Anti-SLC25A1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SAFC
Isopropyl β-D-1-thiogalactopyranoside
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Millipore
Urea solution, suitable for microbiology, 40% in H2O