Merck
  • A method for the isolation and culture of embryonic cardiomyocytes from Mexican axolotl.
A method for the isolation and culture of embryonic cardiomyocytes from Mexican axolotl.
The International journal of developmental biology (1996-08-01)
R W Zajdel, Y Zhu, M E Fransen, L F Lemanski
Sigma-Aldrich
Trypsin solution from porcine pancreas, 1 ×, sterile-filtered, BioReagent, suitable for cell culture, 2.5 g porcine trypsin per liter in Hanks′ Balanced Salt Solution with phenol red