Merck
  • Effect of the method of dispersion on the properties of carbon nanotube and microfibrillar cellulose composites
Effect of the method of dispersion on the properties of carbon nanotube and microfibrillar cellulose composites
Inventia Report (2013)
Nadera A