- Optimum concentration of trifluoroacetic acid for reversed-phase liquid chromatography of peptides revisited.
Optimum concentration of trifluoroacetic acid for reversed-phase liquid chromatography of peptides revisited.
Journal of Chromatography A (2004)
Chen Y, et al.