Merck
  • The effect of combined treatment with risperidone and antidepressants on the MK-801-induced deficits in the social interaction test in rats.
The effect of combined treatment with risperidone and antidepressants on the MK-801-induced deficits in the social interaction test in rats.
Pharmacological reports : PR (2015-10-21)
Katarzyna Kamińska, Zofia Rogóż
ABSTRACT
Several clinical reports have suggested that augmentation of atypical antipsychotics' activity by antidepressants may efficiently improve the treatment of negative and some cognitive symptoms of schizophrenia. The aim of the present study was to investigate the effect of antidepressant mirtazapine or escitalopram and risperidone (an atypical antipsychotic), given separately or jointly, on the MK-801-induced deficits in the social interaction test in rats. Antidepressants and risperidone were given 60 and 30 min before the test, respectively. The social interaction of male Wistar rats was measured for 10 min, starting 4 h after MK-801 (0.1 mg/kg) administration. In the social interaction test, MK-801-induced deficits in the parameters studied, i.e. the number of episodes and the time of interactions. Risperidone at a higher dose (0.1 mg/kg) reversed that effect. Co-treatment with an ineffective dose of risperidone (0.01 mg/kg) and mirtazapine (2.5 or 5 mg/kg) or escitalopram only at a dose of 5 mg/kg (but not 2.5 and 10 mg/kg) abolished the deficits evoked by MK-801. The obtained results suggest that especially mirtazapine, and to a smaller degree escitalopram may enhance the antipsychotic-like effect of risperidone in the animal test modeling some negative symptoms of schizophrenia.
Sigma-Aldrich
Mirtazapine, ≥98% (HPLC)
Sigma-Aldrich
Escitalopram oxalate, ≥98% (HPLC), powder
Sigma-Aldrich
Risperidone, ≥98% (HPLC), powder