Merck
Inhibition of DYRK1A and GSK3B induces human β-cell proliferation.
Nature communications (2015-10-27)
Weijun Shen, Brandon Taylor, Qihui Jin, Van Nguyen-Tran, Shelly Meeusen, You-Qing Zhang, Anwesh Kamireddy, Austin Swafford, Andrew F Powers, John Walker, John Lamb, Badry Bursalaya, Michael DiDonato, George Harb, Minhua Qiu, Christophe M Filippi, Lisa Deaton, Carolina N Turk, Wilma L Suarez-Pinzon, Yahu Liu, Xueshi Hao, Tingting Mo, Shanshan Yan, Jing Li, Ann E Herman, Bernhard J Hering, Tom Wu, H Martin Seidel, Peter McNamara, Richard Glynne, Bryan Laffitte
ABSTRACT
Insufficient pancreatic β-cell mass or function results in diabetes mellitus. While significant progress has been made in regulating insulin secretion from β-cells in diabetic patients, no pharmacological agents have been described that increase β-cell replication in humans. Here we report aminopyrazine compounds that stimulate robust β-cell proliferation in adult primary islets, most likely as a result of combined inhibition of DYRK1A and GSK3B. Aminopyrazine-treated human islets retain functionality in vitro and after transplantation into diabetic mice. Oral dosing of these compounds in diabetic mice induces β-cell proliferation, increases β-cell mass and insulin content, and improves glycaemic control. Biochemical, genetic and cell biology data point to Dyrk1a as the key molecular target. This study supports the feasibility of treating diabetes with an oral therapy to restore β-cell mass, and highlights a tractable pathway for future drug discovery efforts.
Sigma-Aldrich
DAPI, for nucleic acid staining
Pricing and availability is not currently available.