Merck
  • Release of copper from embedded solid copper bullets into muscle and fat tissues of fallow deer (Dama dama), roe deer (Capreolus capreolus), and wild boar (Sus scrofa) and effect of copper content on oxidative stability of heat-processed meat.
Release of copper from embedded solid copper bullets into muscle and fat tissues of fallow deer (Dama dama), roe deer (Capreolus capreolus), and wild boar (Sus scrofa) and effect of copper content on oxidative stability of heat-processed meat.
Meat science (2015-05-26)
I Schuhmann-Irschik, M Sager, P Paulsen, A Tichy, F Bauer
ABSTRACT
When venison with embedded copper bullets was subjected to different culinary processing procedures, the amount of copper released from the embedded bullet was affected more by the retention period of the bullet in the meat during cool storage, than by the different heating protocols. The presence of copper fragments had no significant effect on levels of thiobarbituric acid reactive substances (TBARS). Conversely, TBARS in lean meat (fallow deer, wild boar, roe deer) were significantly affected by culinary treatment (higher TBARS in boiled and boiled-stored meat than in meat barbecued or boiled in brine). In pork-beef patties doped with up to 28mg/kg Cu, TBARS increased after dry-heating and subsequently storing the meat patties. The amount of copper doping had no effect on TBARS for 0 and 7days of storage, but a significant effect at day 14 (fat oxidation retarded at higher Cu doses). Evidence is presented that wild boar meat may be more sensitive to fat oxidation than pork-beef.
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N