Merck
  • The use of dynamic O-(2-18F-fluoroethyl)-l-tyrosine PET in the diagnosis of patients with progressive and recurrent glioma.
The use of dynamic O-(2-18F-fluoroethyl)-l-tyrosine PET in the diagnosis of patients with progressive and recurrent glioma.
Neuro-oncology (2015-05-27)
Norbert Galldiks, Gabriele Stoffels, Christian Filss, Marion Rapp, Tobias Blau, Caroline Tscherpel, Garry Ceccon, Veronika Dunkl, Martin Weinzierl, Michael Stoffel, Michael Sabel, Gereon R Fink, Nadim J Shah, Karl-Josef Langen
ABSTRACT
We evaluated the diagnostic value of static and dynamic O-(2-[(18)F]fluoroethyl)-L-tyrosine ((18)F-FET) PET parameters in patients with progressive or recurrent glioma. We retrospectively analyzed 132 dynamic (18)F-FET PET and conventional MRI scans of 124 glioma patients (primary World Health Organization grade II, n = 55; grade III, n = 19; grade IV, n = 50; mean age, 52 ± 14 y). Patients had been referred for PET assessment with clinical signs and/or MRI findings suggestive of tumor progression or recurrence based on Response Assessment in Neuro-Oncology criteria. Maximum and mean tumor/brain ratios of (18)F-FET uptake were determined (20-40 min post-injection) as well as tracer uptake kinetics (ie, time to peak and patterns of the time-activity curves). Diagnoses were confirmed histologically (95%) or by clinical follow-up (5%). Diagnostic accuracies of PET and MR parameters for the detection of tumor progression or recurrence were evaluated by receiver operating characteristic analyses/chi-square test. Tumor progression or recurrence could be diagnosed in 121 of 132 cases (92%). MRI and (18)F-FET PET findings were concordant in 84% and discordant in 16%. Compared with the diagnostic accuracy of conventional MRI to diagnose tumor progression or recurrence (85%), a higher accuracy (93%) was achieved by (18)F-FET PET when a mean tumor/brain ratio ≥2.0 or time to peak <45 min was present (sensitivity, 93%; specificity, 100%; accuracy, 93%; positive predictive value, 100%; P < .001). Static and dynamic (18)F-FET PET parameters differentiate progressive or recurrent glioma from treatment-related nonneoplastic changes with higher accuracy than conventional MRI.
Product Description
Tantalum(V) ethoxide, packaged for use in deposition systems
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis