Merck
  • MicroRNA-423-5p targets O-GlcNAc transferase to induce apoptosis in cardiomyocytes.
MicroRNA-423-5p targets O-GlcNAc transferase to induce apoptosis in cardiomyocytes.
Molecular medicine reports (2015-03-18)
Peng Luo, Tao He, Rongjian Jiang, Gang Li
ABSTRACT
Congestive heart failure (CHF) remains the single most prevalent cause of mortality and morbidity; therefore, the identification of novel biomarkers is required for the diagnosis of CHF. The aim of the present study was to examine whether microRNA (miR)-423-5p is a novel biomarker for CHF, which was defined by the circulatory biomarker pro-brain natriuretic peptide (BNP). Samples of plasma from patients with CHF and healthy controls were collected and used for quantitative polymerase chain reaction analysis. In addition, the O-GlcNAc transferase (OGT) 3' untranslated region was cloned and used for a luciferase assay. The effects of the expression of miR-423-5p in cardiomyocytes were determined using western blotting and terminal deoxynucleotidyl transferase-deoxyuridine triphosphate nick-end labeling analyses. The results demonstrated that miR-423-5p was associated with CHF and the expression levels of proBNP; in addition, OGT was found to be a direct target of miR-423-5p. The expression of miR-423-5p significantly regulated the expression of OGT and its associated downstream targets and induced apoptosis in the cardiomyocytes. Therefore, the results of the present study indicated that miR-423-5p was involved in CHF via the direct targeting of OGT and the induction of apoptosis in cardiomyocytes.
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Magnesium chloride, ≥98%
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O