Merck
  • A novel fluorescent probe for Au(III)/Au(I) ions based on an intramolecular hydroamination of a Bodipy derivative and its application to bioimaging.
A novel fluorescent probe for Au(III)/Au(I) ions based on an intramolecular hydroamination of a Bodipy derivative and its application to bioimaging.
Chemical communications (Cambridge, England) (2011-11-29)
Jian-Bo Wang, Qing-Qing Wu, Yuan-Zeng Min, Yang-Zhong Liu, Qin-Hua Song
ABSTRACT
A novel fluorescent probe for gold ions (Au(3+)/Au(+)) is reported through blocking photoinduced electron transfer, in which a boron dipyrromethene (Bodipy) derivative reveals high selectivity and sensitivity in a gold-catalyzed intramolecular hydroamination, and is successfully applied to fluorescence imaging of Au(3+) in living cells.
Sigma-Aldrich
4-Bromo-3-nitrobenzaldehyde, 97%
Pricing and availability is not currently available.