Merck
  • Gold nanoparticles enhance the effect of tyrosine kinase inhibitors in acute myeloid leukemia therapy.
Gold nanoparticles enhance the effect of tyrosine kinase inhibitors in acute myeloid leukemia therapy.
International journal of nanomedicine (2016-03-02)
Bobe Petrushev, Sanda Boca, Timea Simon, Cristian Berce, Ioana Frinc, Delia Dima, Sonia Selicean, Grigore-Aristide Gafencu, Alina Tanase, Mihnea Zdrenghea, Adrian Florea, Sorina Suarasan, Liana Dima, Raluca Stanciu, Ancuta Jurj, Anca Buzoianu, Andrei Cucuianu, Simion Astilean, Alexandru Irimie, Ciprian Tomuleasa, Ioana Berindan-Neagoe
ABSTRACT
Every year, in Europe, acute myeloid leukemia (AML) is diagnosed in thousands of adults. For most subtypes of AML, the backbone of treatment was introduced nearly 40 years ago as a combination of cytosine arabinoside with an anthracycline. This therapy is still the worldwide standard of care. Two-thirds of patients achieve complete remission, although most of them ultimately relapse. Since the FLT3 mutation is the most frequent, it serves as a key molecular target for tyrosine kinase inhibitors (TKIs) that inhibit FLT3 kinase. In this study, we report the conjugation of TKIs onto spherical gold nanoparticles. The internalization of TKI-nanocarriers was proved by the strongly scattered light from gold nanoparticles and was correlated with the results obtained by transmission electron microscopy and dark-field microscopy. The therapeutic effect of the newly designed drugs was investigated by several methods including cell counting assay as well as the MTT assay. We report the newly described bioconjugates to be superior when compared with the drug alone, with data confirmed by state-of-the-art analyses of internalization, cell biology, gene analysis for FLT3-IDT gene, and Western blotting to assess degradation of the FLT3 protein. The effective transmembrane delivery and increased efficacy validate its use as a potential therapeutic.
Sigma-Aldrich
CEP-701 hydrate, ≥98% (HPLC)
Pricing and availability is not currently available.