Merck
  • Structure-Based Consensus Scoring Scheme for Selecting Class A Aminergic GPCR Fragments.
Structure-Based Consensus Scoring Scheme for Selecting Class A Aminergic GPCR Fragments.
Journal of chemical information and modeling (2016-01-14)
Ádám A Kelemen, Róbert Kiss, György G Ferenczy, László Kovács, Beáta Flachner, Zsolt Lőrincz, György M Keserű
ABSTRACT
Aminergic G-protein coupled receptors (GPRCs) represent well-known targets of central nervous-system related diseases. In this study a structure-based consensus virtual screening scheme was developed for designing targeted fragment libraries against class A aminergic GPCRs. Nine representative aminergic GPCR structures were selected by first clustering available X-ray structures and then choosing the one in each cluster that performs best in self-docking calculations. A consensus scoring protocol was developed using known promiscuous aminergic ligands and decoys as a training set. The consensus score (FrACS-fragment aminergic consensus score) calculated for the optimized protein ensemble showed improved enrichments in most cases as compared to stand-alone structures. Retrospective validation was carried out on public screening data for aminergic targets (5-HT1 serotonin receptor, TA1 trace-amine receptor) showing 8-17-fold enrichments using an ensemble of aminergic receptor structures. The performance of the structure based FrACS in combination with our ligand-based prefilter (FrAGS) was investigated both in a retrospective validation on the ChEMBL database and in a prospective validation on an in-house fragment library. In prospective validation virtual fragment hits were tested on 5-HT6 serotonin receptors not involved in the development of FrACS. Six out of the 36 experimentally tested fragments exhibited remarkable antagonist efficacies, and 4 showed IC50 values in the low micromolar or submicromolar range in a cell-based assay. Both retrospective and prospective validations revealed that the methodology is suitable for designing focused class A GPCR fragment libraries from large screening decks, commercial compound collections, or virtual databases.
Sigma-Aldrich
Maltose solution, BioReagent, ~20% in H2O, Molecular Biology