Merck
  • Synthesis and physical properties of estolide ester using saturated Fatty Acid and ricinoleic Acid.
Synthesis and physical properties of estolide ester using saturated Fatty Acid and ricinoleic Acid.
Journal of automated methods & management in chemistry (2011-10-19)
Jumat Salimon, Neeranjini Nallathamby, Nadia Salih, Bashar Mudhaffar Abdullah
ABSTRACT
A study was conveyed to produce estolide ester using ricinoleic acid as the backbone. The ricinoleic acid reacted with saturated fatty acid from C8-C18. These reactions were conducted under vacuum at 60°C for 24 h without solvent. The reaction used acid catalyst, sulphuric acid. The new saturate ricinoleic estolide esters show superior low-temperature properties (-52 ± 0.08°C) and high flash point (>300°C). The yield of the neat estolide esters ranged from 52% to 96%. The viscosity range was 51 ± 0.08 to 86 ± 0.01 cp. These new saturated estolide esters were also compared with saturated branched estolide esters.
Sigma-Aldrich
2-Butyloctanoic acid, 96%
Pricing and availability is not currently available.