Merck
  • Preparation and evaluation of hybrid composites of chemical fuel and multi-walled carbon nanotubes in the study of thermopower waves.
Preparation and evaluation of hybrid composites of chemical fuel and multi-walled carbon nanotubes in the study of thermopower waves.
Journal of Visualized Experiments (2015)
Hwang H, et al.