- A Caenorhabditis elegans locomotion phenotype caused by transgenic repeats of the hlh-17 promoter sequence.
A Caenorhabditis elegans locomotion phenotype caused by transgenic repeats of the hlh-17 promoter sequence.
PloS one (2013-12-07)
Randy F Stout, Vladimir Grubišić, Vladimir Parpura
PMID24312354
ABSTRACT
Transgene technology is one of the most heavily relied upon tools in modern biological research. Expression of an exogenous gene within cells, for research and therapeutic applications, nearly always includes promoters and other regulatory sequences. We found that repeats of a non-protein coding transgenic sequence produced profound changes to the behavior of the nematode Caenorhabditis elegans. These changes were produced by a glial promoter sequence but, unexpectedly, major deficits were observed specifically in backward locomotion, a neuron-driven behavior. We also present evidence that this behavioral phenotype is transpromoter copy number-dependent and manifests early in development and is maintained into adulthood of the worm.
Product Description
Roche
FastStart™ PCR Master, sufficient for ≤100 reactions (04710436001), sufficient for ≤400 reactions (04710444001), sufficient for ≤2,000 reactions (04710452001)
Pricing and availability is not currently available.