Merck
  • Transfected human liver cytochrome P-450 hydroxylates vitamin D analogs at different side-chain positions.
Transfected human liver cytochrome P-450 hydroxylates vitamin D analogs at different side-chain positions.
Proceedings of the National Academy of Sciences of the United States of America (1993-09-15)
Y D Guo, S Strugnell, D W Back, G Jones
ABSTRACT
A full-length cDNA for the human liver mitochondrial cytochrome P-450 CYP27 was cloned from a human hepatoma HepG2 cDNA library and then subcloned into the mammalian expression vector pSG5. When CYP27 cDNA was transfected into COS-1 transformed monkey kidney cells along with adrenodoxin cDNA, transfected cells revealed a 10- to 20-fold higher vitamin D3-25-hydroxylase activity than nontransfected cells. Transfected cells were capable of 25-hydroxylation of vitamin D3, 1 alpha-hydroxyvitamin D3 and 1 alpha-hydroxydihydrotachysterol3. In each case they also showed the ability to 26(27)-hydroxylate the cholesterol-like (D3) side chain. The relative rates of 25- and 26(27)-hydroxylation of 1 alpha-hydroxyvitamin D3 approximately mimicked the ratio of products observed in HepG2 cells. Vitamin D2 and 1 alpha-hydroxyvitamin D2, both with the ergosterol-like side chain, were 24- and 26(27)-hydroxylated by CYP27. The rate of side-chain 24-, 25-, or 26(27)-hydroxylation was greater for 1 alpha-hydroxylated vitamin D analogs than for their nonhydroxylated counterparts. We conclude that CYP27 is capable of 24-, 25-, and 26(27)-hydroxylation of vitamin D analogs and that the nature of products is partially dictated by the side chain of the substrate. This work has revealed that the cytochrome P-450 CYP27 may be important in the metabolism of vitamin D analogs used as drugs.