Merck
Developments in biotechnological production of sweet proteins.
Journal of bioscience and bioengineering (2006-12-26)
Tetsuya Masuda, Naofumi Kitabatake
ABSTRACT
Most proteins are tasteless and flavorless, while some proteins elicit a sweet-taste response on the human palate. Six proteins, thaumatin, monellin, mabinlin, brazzein, egg lysozyme, and neoculin (previously considered as curculin) have been identified as sweet-tasting proteins. However, no common features among them have been observed. Herein, recent advances in the research of sweet-tasting proteins and the production of such proteins by biotechnological approaches are reviewed. Information on the structure-sweetness relationship for these proteins would help not only in the clarification of the mechanism of interaction of sweet-tasting proteins with their receptors, but also in the design of more effective low-calorie sweeteners.
Sigma-Aldrich
Thaumatin from Thaumatococcus daniellii
Pricing and availability is not currently available.