Merck
  • Enhanced expression of Fas and FasL modulates apoptosis in the lungs of severe P. falciparum malaria patients with pulmonary edema.
Enhanced expression of Fas and FasL modulates apoptosis in the lungs of severe P. falciparum malaria patients with pulmonary edema.
International Journal of Clinical and Experimental Pathology (2015)
Punsawad C, et al.