Merck
  • Hypoxia-induced expression of cellular prion protein improves the therapeutic potential of mesenchymal stem cells.
Hypoxia-induced expression of cellular prion protein improves the therapeutic potential of mesenchymal stem cells.
Cell death & disease (2016-10-07)
Yong-Seok Han, Jun Hee Lee, Yeo Min Yoon, Chul Won Yun, Hyunjin Noh, Sang Hun Lee
ABSTRACT
Mesenchymal stem cells (MSCs) are 'adult' multipotent cells that promote regeneration of injured tissues in vivo. However, differences in oxygenation levels between normoxic culture conditions (21% oxygen) and both the MSC niche (2-8% oxygen) and ischemic injury-induced oxidative stress conditions in vivo have resulted in low efficacy of MSC therapies in both pre-clinical and clinical studies. To address this issue, we examined the effectiveness of hypoxia preconditioning (2% oxygen) for enhancing the bioactivity and tissue-regenerative potential of adipose-derived MSCs. Hypoxia preconditioning enhanced the proliferative potential of MSCs by promoting the expression of normal cellular prion protein (PrP
Sigma-Aldrich
MISSION® esiRNA, targeting human PRNP
Pricing and availability is not currently available.