Merck
  • period-1 encodes an ATP-dependent RNA helicase that influences nutritional compensation of the Neurospora circadian clock.
period-1 encodes an ATP-dependent RNA helicase that influences nutritional compensation of the Neurospora circadian clock.
Proceedings of the National Academy of Sciences of the USA (2015)
Emerson JM, et al.